Få mere at vide
3,5,6-Trihydroxy-1-isopropylindole, TRC
Artikelnummer. 30451124
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30451124 25 mg -
30450808 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451124 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79528525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 73853-05-1
Molekylær formel C11 H13 N O3
Molekylvægt (g/mol) 207.226
Synonym 1-(1-Methylethyl)-1H-indole-3,5,6-triol
IUPAC navn 1-propan-2-ylindole-3,5,6-triol
SMIL CC(C)n1cc(O)c2cc(O)c(O)cc12
Tekniske data
Kemisk navn eller materiale 3,5,6-Trihydroxy-1-isopropylindole
Mængde 25 mg
InChI formel InChI=1S/C11H13NO3/c1-6(2)12-5-11(15)7-3-9(13)10(14)4-8(7)12/h3-6,13-15H,1-2H3
Formel vægt 207.09