Få mere at vide
3,5,3',5'-Tetrachlorobisphenol A, TRC
Artikelnummer. 17980875
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17980875 100 mg
17964115 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17980875 Leverandør Toronto Research Chemicals Leverandørnr. TRCT291360100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79-95-8
Molekylær formel C15 H12 Cl4 O2
Molekylvægt (g/mol) 366.07
Synonym Phenol, 4,4'-(1-methylethylidene)bis[2,6-dichloro-, Phenol, 4,4'-isopropylidenebis[2,6-dichloro- (6CI,8CI), 4,4'-(1-Methylethylidene)bis[2,6-dichlorophenol], 2,2-Bis(3,5-dichloro-4-hydroxyphenyl)propane, 2,2-Bis(4-hydroxy-3,5-dichlorophenyl)propane, 2,2-Bis[3,5-dichloro-4-oxyphenyl]propane, 2,2',6,6'-Tetrachlorobisphenol A, 3,3',5,5'-Tetrachlorobisphenol A, 3,5,3',5'-Tetrachlorobisphenol A, 4,4'-Isopropylidenebis[2,6-dichlorophenol], NSC 18248, NSC 67465, Tetrachlorobisphenol A, Tetrachlorodian
IUPAC navn 2,6-dichloro-4-[2-(3,5-dichloro-4-hydroxyphenyl)propan-2-yl]phenol
SMIL CC(C)(c1cc(Cl)c(O)c(Cl)c1)c2cc(Cl)c(O)c(Cl)c2
Tekniske data
Kemisk navn eller materiale Tetrachlorobisphenol A
Mængde 100 mg
InChI formel InChI=1S/C15H12Cl4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21H,1-2H3
Formel vægt 363.9591