missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Pyridyl)-L-alanine dihydrochloride, 97%, Thermo Scientific™

Catalog Number p-7055366
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
178933-04-5
C8H12Cl2N2O2
239.10
MFCD13184932
NAMMTAFKUFJMSD-UHFFFAOYNA-N
s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-4-pyridyl-l-alanine dihydrochloride, 2s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-pyridin-4-yl-l-alanine-hydrogen chloride 1/2, s-2-amino-3-pyridin-4-yl propanoic acid 2hcl, s-2-amino-3-pyridin-4-yl propanoicaciddihydrochloride
20672030
Cl.Cl.NC(CC1=CC=NC=C1)C(O)=O
This item is not returnable. View return policy
178933-04-5
C8H12Cl2N2O2
239.10
MFCD13184932
NAMMTAFKUFJMSD-UHFFFAOYNA-N
s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-4-pyridyl-l-alanine dihydrochloride, 2s-2-amino-3-pyridin-4-yl propanoic acid dihydrochloride, 3-pyridin-4-yl-l-alanine-hydrogen chloride 1/2, s-2-amino-3-pyridin-4-yl propanoic acid 2hcl, s-2-amino-3-pyridin-4-yl propanoicaciddihydrochloride
20672030
Cl.Cl.NC(CC1=CC=NC=C1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
IUPAC Name (2S)-2-amino-3-pyridin-4-ylpropanoic acid;dihydrochloride
Formula Weight 239.1
Percent Purity 97%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material 3-(4-Pyridyl)-L-alanine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'