missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Nitrophenyl)propanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7077722
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
16642-79-8
C9H9NO4
195.17
MFCD00126834
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
85526
3-(4-nitrophenyl)propanoic acid
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O
This item is not returnable. View return policy
16642-79-8
C9H9NO4
195.17
MFCD00126834
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
85526
3-(4-nitrophenyl)propanoic acid
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 166.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 195.18
Percent Purity 98%
Chemical Name or Material 3-(4-Nitrophenyl)propanoic acid
compliance-icons
Product Identifier
  • 3-(4-Nitrophenyl)propanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'