missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Methylthio)phenyl]propionic acid, 96%, Alfa Aesar™

Brand:  Alfa Aesar H34101.09

Additional Details : CAS Number : 138485-81-1 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15437017

  • 1009.00 DKK / 10g

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

138485-81-1
196.264
ASTVFRXZLBGVRL-UHFFFAOYSA-N
3801098
CSC1=CC=C(C=C1)CCC(=O)O
C10H12O2S
MFCD03425696
3-4-methylsulfanyl phenyl propanoic acid, 3-4-methylthio phenyl propanoic acid, 3-4-methylthiophenyl propionic acid, benzenepropanoicacid, 4-methylthio, benzenepropanoic acid, 4-methylthio, 3-4-methylthio phenyl propionic acid, 3-4-methylsulfanylphenyl propanoic acid, acmc-20ex3q, benzenepropanoicacid,4-methylthio
3-(4-methylsulfanylphenyl)propanoic acid

Specifications

3-[4-(Methylthio)phenyl]propionic acid
10g
C10H12O2S
3-4-methylsulfanyl phenyl propanoic acid, 3-4-methylthio phenyl propanoic acid, 3-4-methylthiophenyl propionic acid, benzenepropanoicacid, 4-methylthio, benzenepropanoic acid, 4-methylthio, 3-4-methylthio phenyl propionic acid, 3-4-methylsulfanylphenyl propanoic acid, acmc-20ex3q, benzenepropanoicacid,4-methylthio
CSC1=CC=C(C=C1)CCC(=O)O
196.264
196.26
97°C to 102°C
138485-81-1
MFCD03425696
ASTVFRXZLBGVRL-UHFFFAOYSA-N
3-(4-methylsulfanylphenyl)propanoic acid
3801098
96%
Videos
SDS Documents