Få mere at vide
3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline-d3, TRC
Artikelnummer. 30475801
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475801 50 mg
30469024 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475801 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31306750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246815-34-8
Molekylær formel C11H7D3F3N3
Molekylvægt (g/mol) 244.24
Synonym 3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)benzenamine-d3, 1-(3-Amino-5-trifluoromethylphenyl)-4-methylimidazole-d3
IUPAC navn 3-[4-(trideuteriomethyl)imidazol-1-yl]-5-(trifluoromethyl)aniline
SMIL [2H]C([2H])([2H])c1cn(cn1)c2cc(N)cc(c2)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline-d3
Mængde 50 mg
InChI formel InChI=1S/C11H10F3N3/c1-7-5-17(6-16-7)10-3-8(11(12,13)14)2-9(15)4-10/h2-6H,15H2,1H3/i1D3
Formel vægt 244.1015