Få mere at vide
3-(4-Hydroxyphenyl)-pyrazin-2-ol, Mikromol™
Artikelnummer. 30049811
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049811 25 mg
30049240 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049811 Leverandør Mikromol Leverandørnr. MM0232.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(4-Hydroxyphenyl)-pyrazin-2-ol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(4-Hydroxyphenyl)-pyrazin-2-ol
CAS 126247-63-0
Analyt- eller komponentnavne 3-(4-Hydroxyphenyl)-pyrazin-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C10 H8 N2 O2
Certificeringer/compliance ISO 17025
Synonym Amoxicillin Trihydrate Imp. F (EP), Amoxicillin Imp. F (EP), Amoxicillin USP Related Compound F, Amoxicillin USP RC F, 3-(4-Hydroxyphenyl)-pyrazin-2-ol, Ph Eur Amoxicillin Impurity F, Amoxycillin related, SB 258740, Amoxicillin Related Compound F, Amoxicillin Trihydrate Impurity F, Amoxicillin Impurity F
InChI formel InChI=1S/C10H8N2O2/c13-8-3-1-7(2-4-8)9-10(14)12-6-5-11-9/h1-6,13H,(H,12,14)
SMIL Oc1ccc(cc1)c2nccnc2O
IUPAC navn 3-(4-hydroxyphenyl)pyrazin-2-ol
Molekylvægt (g/mol) 188.18
Mængde 25 mg
Formel vægt 188.0586 g/mol