missing translation for 'onlineSavingsMsg'
Få mere at vide

3-(4-Hydroxyphenyl)-1-propanol, TRC

Artikelnummer. 30392799
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30392799 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30392799 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95283025G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-(4-Hydroxyphenyl)-1-propanol
CAS 10210-17-0
Molekylær formel C9H12O2
Mængde 25 g
Synonym 1-(4-Hydroxyphenyl)-3-propanol, 3-(4-Hydroxyphenyl)-1-propanol, 3-(4-Hydroxyphenyl)propanol, 3-(p-Hydroxyphenyl)-1-propanol, 3-(p-Hydroxyphenyl)propyl alcohol, 4-(3-Hydroxypropyl)phenol, 4-Hydroxybenzenepropanol, Dihydro-p-coumaryl Alcohol
InChI formel InChI=1S/C9H12O2/c10-7-1-2-8-3-5-9(11)6-4-8/h3-6,10-11H,1-2,7H2
SMIL OCCCc1ccc(O)cc1
IUPAC navn 4-(3-hydroxypropyl)phenol
Molekylvægt (g/mol) 152.19
Formel vægt 152.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.