missing translation for 'onlineSavingsMsg'
Learn More

3,4-Hexanedione, 94%, Thermo Scientific Chemicals

Catalog Number p-7044474
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
4437-51-8
C6H10O2
114.144
MFCD00010237
KVFQMAZOBTXCAZ-UHFFFAOYSA-N
3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
62539
hexane-3,4-dione
CCC(=O)C(=O)CC
This item is not returnable. View return policy
4437-51-8
C6H10O2
114.144
MFCD00010237
KVFQMAZOBTXCAZ-UHFFFAOYSA-N
3,4-hexanedione, bipropionyl, dipropionyl, diethyl diketone, 3,4-hexane dione, unii-4m4npr5vgd, 3,4-hexandione, fema no. 3168, ccris 6295, 4m4npr5vgd
62539
hexane-3,4-dione
CCC(=O)C(=O)CC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -10°C
Density 0.941
Boiling Point 129°C to 131°C
Flash Point 29°C (84°F)
Refractive Index 1.412
Quantity 25 g
UN Number UN1224
Beilstein 1700837
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material 3,4-Hexanedione
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'