missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Formylphenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-7032422
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
34961-64-3
C10H10O3
178.19
MFCD08234824
NRCCSDVEUWXOMG-UHFFFAOYSA-N
3-4-formylphenyl propanoic acid, 3-4-formylphenyl propionic acid, 3-4-formyl-phenyl-propionic acid, 3-4-formylphenyl-propionic acid, 4-formyldihydrocinnamic acid, 4-formylbenzenepropanoic acid, benzenepropanoic acid,4-formyl, 3-4-formylphenyl propanoicacid
19825241
3-(4-formylphenyl)propanoic acid
OC(=O)CCC1=CC=C(C=O)C=C1
This item is not returnable. View return policy
34961-64-3
C10H10O3
178.19
MFCD08234824
NRCCSDVEUWXOMG-UHFFFAOYSA-N
3-4-formylphenyl propanoic acid, 3-4-formylphenyl propionic acid, 3-4-formyl-phenyl-propionic acid, 3-4-formylphenyl-propionic acid, 4-formyldihydrocinnamic acid, 4-formylbenzenepropanoic acid, benzenepropanoic acid,4-formyl, 3-4-formylphenyl propanoicacid
19825241
3-(4-formylphenyl)propanoic acid
OC(=O)CCC1=CC=C(C=O)C=C1

3-(4-Formylphenyl)propionic acid is used as an intermediate in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Formylphenyl)propionic acid is used as an intermediate in the manufacture of pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼136°C (decomposition)
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 178.19
Percent Purity 96%
Chemical Name or Material 3-(4-Formylphenyl)propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'