Få mere at vide
3,4-Dimethylphenol-2,5,6-d3,OD, CDN
Artikelnummer. 30316488
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316488 0.25 g -
30327183 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316488 Leverandør CDN Leverandørnr. CDND53680.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dimethylphenol-O,2,5,6-D4
CAS 1219803-55-0
Molekylær formel C8 D4 H6 O
Mængde 0.25 g
Synonym 3,4-Dimethylphenol-d4, 3,4-Dimethylphenol-2,5,6-d3,OD, 3,4-Dimethylphenol-O,2,5,6-D4, 3,4-Dimethylphenol-(ring-d3,O-d)
InChI formel InChI=1 S/C8H10O/c1-6-3-4-8(9)5-7(6)2/h3-5,9 H,1-2H3/i3D,4 D,5 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c(C)c(C)c1[2 H]
IUPAC navn 1,2,4-trideuterio-3-deuteriooxy-5,6-dimethylbenzene
Molekylvægt (g/mol) 126.1891
Formel vægt 126.0983 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity