missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-3778324
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5763-61-1
C9H13NO2
167.21
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=CC=C(CN)C=C1OC
This item is not returnable. View return policy
5763-61-1
C9H13NO2
167.21
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=CC=C(CN)C=C1OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 281°C to 284°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2NH2
Refractive Index 1.5530 to 1.557
Quantity 5 g
Beilstein 13, 796
Specific Gravity 1.1
Formula Weight 167.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'