Få mere at vide
3,4-Dimethoxybenzaldehyde-d3, TRC
Artikelnummer. 30372146
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30372146 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372146 Leverandør Toronto Research Chemicals Leverandørnr. TRCD469637250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dimethoxybenzaldehyde-d3
CAS 143318-06-3
Renhedsgrad noter HPLC
Molekylær formel C9 2H3 H7 O3
Mængde 250 mg
Synonym Benzaldehyde, 3-methoxy-4-(methoxy-d3)-, 3-Methoxy-4-(methoxy-d3)benzaldehyde, 3,4-Dimethoxybenzaldehyde-d3, 3,4-Dimethoxybenzaldehyde-D3 (4-methoxy D3), Veratraldehyde-d3, 4-O-(Methyl-d3)vanillin, Vanillin methyl-d3 ether, Benzaldehyde, 3-methoxy-4-(methoxy-d3)-, 3-Methoxy-4-(methoxy-d3)benzaldehyde, 3,4-Dimethoxybenzaldehyde-d3, 3,4-Dimethoxybenzaldehyde-D3 (4-methoxy-D3), Veratraldehyde-d3, 4-O-(Methyl-d3)vanillin, Vanillin methyl-d3 ether
InChI formel InChI=1S/C9H10O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-6H,1-2H3/i1D3
SMIL [2H]C([2H])([2H])Oc1ccc(C=O)cc1OC
IUPAC navn 3-methoxy-4-(trideuteriomethoxy)benzaldehyde
Molekylvægt (g/mol) 169.19
Formel vægt 169.0818
Procent renhed >95