Få mere at vide
3',4'-Dimethoxyaceto-d3-phenone, CDN
Artikelnummer. 30313787
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313787 0.5 g
30313571 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313787 Leverandør CDN Leverandørnr. CDND50370.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dimethoxyacetophenone-D3 (acetyl-D3)
CAS 350818-54-1
Molekylær formel C10 D3 H9 O3
Mængde 0.5 g
Synonym Ethanone-2,2,2-d3, 1-(3,4-dimethoxyphenyl)- (9 CI), 1-(3,4-Dimethoxyphenyl)ethanone-2,2,2-d3, 3,4-Dimethoxyacetophenone-d3, 3,4-Dimethoxyacetophenone-D3 (acetyl-D3), 3,4-Dimethoxyphenyl methyl-d3 ketone, 3',4'-Dimethoxyacetophenone-d3, 3',4'-Dimethoxyacetophenone-d3, 3',4'-Dimethoxyacetophenone-d3, 3',4'-Dimethoxyacetophenone-d3
InChI formel InChI=1 S/C10H12O3/c1-7(11)8-4-5-9(12-2)10(6-8)13-3/h4-6 H,1-3H3/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)c1ccc(OC)c(OC)c1
IUPAC navn 2,2,2-trideuterio-1-(3,4-dimethoxyphenyl)ethanone
Molekylvægt (g/mol) 183.219
Formel vægt 183.097 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity