Få mere at vide
3,4-Dihydroxyphenylacetic Acid-d5, TRC
Artikelnummer. 30376439
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30376439 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376439 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45425310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dihydroxyphenylacetic Acid-d5
CAS 60696-39-1
Renhedsgrad noter HPLC
Molekylær formel C8 D5 H3 O4
Mængde 10 mg
Synonym Benzene-2,3,6-d3-acetic-α,α-d2 acid, 4,5-dihydroxy- (9CI), 3,4-Dihydroxybenzene-2,5,6-d3-acetic-α,α-d2 acid, (3,4-Dihydroxyphenyl-2,5,6-d3)acetic-d2 acid, 2-(3,4-Dihydroxyphenyl-2,5,6-d3)acetic-2,2-d2 acid, 3',4'-Dihydroxyphenylacetic-d5 acid, 3',4'-Dihydroxyphenylacetic Acid-D5, 3',4'-Dihydroxyphenylacetic acid-2,2,2',5',6'-d5
InChI formel InChI=1S/C8H8O4/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)/i1D,2D,3D,4D2
SMIL [2H]c1c([2H])c(c([2H])c(O)c1O)C([2H])([2H])C(=O)O
IUPAC navn 2,2-dideuterio-2-(2,3,6-trideuterio-4,5-dihydroxyphenyl)acetic acid
Molekylvægt (g/mol) 173.178
Formel vægt 173.074
Procent renhed >95