Få mere at vide
(3,4-Dihydroxyphenyl-d3)acetic-2,2-d2 Acid, CDN
Artikelnummer. 30320068
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320068 0.1 g 0.1g
30319139 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320068 Leverandør CDN Leverandørnr. CDND16420.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dihydroxyphenylacetic Acid-d5
CAS 60696-39-1
Molekylær formel C8 D5 H3 O4
Mængde 0.1 g
Synonym Benzene-2,3,6-d3-acetic-α,α-d2 acid, 4,5-dihydroxy- (9 CI), 3,4-Dihydroxybenzene-2,5,6-d3-acetic-α,α-d2 acid, (3,4-Dihydroxyphenyl-2,5,6-d3)acetic-d2 acid, 2-(3,4-Dihydroxyphenyl-2,5,6-d3)acetic-2,2-d2 acid, 3',4'-Dihydroxyphenylacetic-d5 acid, 3',4'-Dihydroxyphenylacetic acid-2,2,2',5',6'-d5, 3',4'-Dihydroxyphenylacetic acid-2,2,2',5',6'-d5
InChI formel InChI=1 S/C8H8O4/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3,9-10 H,4H2,(H,11,12)/i1D,2 D,3 D,4D2
SMIL [2 H]c1c([2 H])c(c([2 H])c(O)c1O)C([2 H])([2 H])C(=O)O
IUPAC navn 2,2-dideuterio-2-(2,3,6-trideuterio-4,5-dihydroxyphenyl)acetic acid
Molekylvægt (g/mol) 173.178
Formel vægt 173.074 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity