Få mere at vide
3,4-Dihydroxy Styrene (stabilized with BHT 1 % w/w), TRC
Artikelnummer. 30135906
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135906 25 mg
30133530 5 mg
30011121 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135906 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45480525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6053-02-7
Molekylær formel C8 H8 O2
Molekylvægt (g/mol) 136.15
Synonym 4-Ethenyl-1,2-benzenediol, 3,4-Styrenediol, 4-Vinylcatechol, 4-Vinylpyrocatechol, Vinylcatechol;
IUPAC navn 4-ethenylbenzene-1,2-diol
SMIL Oc1ccc(C=C)cc1O
Tekniske data
Kemisk navn eller materiale 3,4-Dihydroxy Styrene
Mængde 25 mg
InChI formel InChI=1S/C8H8O2/c1-2-6-3-4-7(9)8(10)5-6/h2-5,9-10H,1H2
Formel vægt 136.0524