Få mere at vide
3,4-Dihydroagomelatine, TRC
Artikelnummer. 30134044
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134044 50 mg
30133722 5 mg
30007375 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134044 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44825550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1352139-51-5
Molekylær formel C15H19NO2
Molekylvægt (g/mol) 245.32
Synonym N-[2-(3,4-Dihydro-7-methoxy-1-naphthyl)ethyl]acetamide, Agomelatine Impurity 1;
SMIL COc1ccc2CCC=C(CCNC(=O)C)c2c1
Tekniske data
Kemisk navn eller materiale 3,4-Dihydroagomelatine
Mængde 50 mg
InChI formel InChI=1S/C15H19NO2/c1-11(17)16-9-8-13-5-3-4-12-6-7-14(18-2)10-15(12)13/h5-7,10H,3-4,8-9H2,1-2H3,(H,16,17)
Formel vægt 245.1416