Få mere at vide
3,4-Dihydro-7-(4-hydroxybutoxy)-2(1H)-quinolinone, TRC
Artikelnummer. 17974765
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17974765 10 g
17961935 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17974765 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44954010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 889443-20-3
Molekylær formel C13 H17 N O3
Molekylvægt (g/mol) 235.28
Synonym 2(1H)-Quinolinone, 3,4-dihydro-7-(4-hydroxybutoxy)-, 7-(4-Hydroxybutoxy)-3,4-dihydroquinolin-2(1H)-one, 3,4-Dihydro-7-(4-hydroxybutoxy)-2(1H)-quinolinone, Aripiprazole Related Compound B
IUPAC navn 7-(4-hydroxybutoxy)-3,4-dihydro-1H-quinolin-2-one
SMIL OCCCCOc1ccc2CCC(=O)Nc2c1
Tekniske data
Kemisk navn eller materiale 3,4-Dihydro-7-(4-hydroxybutoxy)-2(1H)-quinolinone
Mængde 10 g
InChI formel InChI=1S/C13H17NO3/c15-7-1-2-8-17-11-5-3-10-4-6-13(16)14-12(10)9-11/h3,5,9,15H,1-2,4,6-8H2,(H,14,16)
Formel vægt 235.1208