missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran-5-carbaldehyde, Thermo Scientific™

Catalog Number p-2699403
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
25090-33-9
C6H8O2
112.128
MFCD00085019
WWZVSCJTMPYTLY-UHFFFAOYSA-N
2736151
3,4-dihydro-2H-pyran-5-carbaldehyde
C1CC(=COC1)C=O
This item is not returnable. View return policy
25090-33-9
C6H8O2
112.128
MFCD00085019
WWZVSCJTMPYTLY-UHFFFAOYSA-N
2736151
3,4-dihydro-2H-pyran-5-carbaldehyde
C1CC(=COC1)C=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 5,6-dihydro-4h-pyran-3-carbaldehyde, 2h-pyran-5-carboxaldehyde,3,4-dihydro, 2h-pyran-5-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-pyran-5-carbaldehyd, 3,4-dihydro-2h-pyran-5-carboxaldehyde, 3,4-dihydro-2h-pyrane-5-carbaldehyde
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-pyran-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'