missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-N-(4-fluorobenzyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog Number p-7041330
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
321705-45-7
C13H10F3NO2S
301.28
MFCD02556043
AFXHLFLQECWKRV-UHFFFAOYSA-N
7790813
3,4-difluoro-N-[(4-fluorophenyl)methyl]benzenesulfonamide
FC1=CC=C(CNS(=O)(=O)C2=CC(F)=C(F)C=C2)C=C1
This item is not returnable. View return policy
321705-45-7
C13H10F3NO2S
301.28
MFCD02556043
AFXHLFLQECWKRV-UHFFFAOYSA-N
7790813
3,4-difluoro-N-[(4-fluorophenyl)methyl]benzenesulfonamide
FC1=CC=C(CNS(=O)(=O)C2=CC(F)=C(F)C=C2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 301.28
Percent Purity 97%
Chemical Name or Material 3,4-Difluoro-N-(4-fluorobenzyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'