missing translation for 'onlineSavingsMsg'
Få mere at vide

3',4'-dichlorpropiofenon, TRC

Artikelnummer. 30376484
Change view
Click to view available options
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30376484 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376484 Supplier Toronto Research Chemicals Supplier No. TRCD43627010G
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3',4'-Dichloropropiophenone
CAS 6582-42-9
Molekylær formel C9 H8 Cl2 O
Mængde 10 g
Synonym 1-(3,4-Dichlorophenyl)-1-propanone, 3',4'-Dichloropropiophenone
InChI formel InChI=1S/C9H8Cl2O/c1-2-9(12)6-3-4-7(10)8(11)5-6/h3-5H,2H2,1H3
SMIL CCC(=O)c1ccc(Cl)c(Cl)c1
IUPAC navn 1-(3,4-dichlorophenyl)propan-1-one
Molekylvægt (g/mol) 203.065
Formel vægt 201.995
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.