missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
3,4'-Dichlorodiphenyl Ether, Mikromol™
Beskrivelse
3,4'-Dichlorodiphenyl Ether
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3,4'-Dichlorodiphenyl Ether |
| CAS | 6842-62-2 |
| Analyt- eller komponentnavne | 3,4'-Dichlorodiphenyl Ether |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H8 Cl2 O |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Benzene, 1-chloro-3-(4-chlorophenoxy)-, Ether, m-chlorophenyl p-chlorophenyl (7CI,8CI), 3,4'-Dichlorodiphenyl ether, PCDE 13 |
| InChI formel | InChI=1S/C12H8Cl2O/c13-9-4-6-11(7-5-9)15-12-3-1-2-10(14)8-12/h1-8H |
| SMIL | Clc1ccc(Oc2cccc(Cl)c2)cc1 |
| IUPAC navn | 1-chloro-3-(4-chlorophenoxy)benzene |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?