Learn More
3,4-Dichlorobenzophenone, TRC
Product Code. 30377501
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30377501 5 g 5g
1 options
Product Code. 30377501 Supplier Toronto Research Chemicals Supplier No. TRCD4319655G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,4-Dichlorobenzophenone
CAS 6284-79-3
Molecular Formula C13H8Cl2O
Quantity 5 g
Synonym Benzophenone, 3,4-dichloro- (6CI,8CI), (3,4-Dichlorophenyl)phenylmethanone, (3,4-Dichlorophenyl)(phenyl)methanone, 3,4-Dichlorophenyl phenyl ketone, NSC 5249, Phenyl 3,4-dichlorophenyl ketone
InChI Formula InChI=1S/C13H8Cl2O/c14-11-7-6-10(8-12(11)15)13(16)9-4-2-1-3-5-9/h1-8H
SMILES Clc1ccc(cc1Cl)C(=O)c2ccccc2
IUPAC Name (3,4-dichlorophenyl)-phenylmethanone
Molecular Weight (g/mol) 251.11
Formula Weight 249.9952