Få mere at vide
3,4-Dichlorobenzoic-d3 Acid, CDN
Artikelnummer. 30327924
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327924 0.25 g -
30311860 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327924 Leverandør CDN Leverandørnr. CDND50310.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dichlorobenzoic Acid-D3
CAS 350818-53-0
Molekylær formel C7 D3 H Cl2 O2
Mængde 0.25 g
Synonym Benzoic-2,3,6-d3 acid, 4,5-dichloro- (9 CI), 3,4-Dichlorobenzoic-d3 Acid, 3,4-Dichlorobenzoic Acid-D3, 3,4-Dichlorobenzoic-2,5,6-d3 acid
InChI formel InChI=1 S/C7H4Cl2O2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3 H,(H,10,11)/i1D,2 D,3 D
SMIL [2 H]c1c([2 H])c(C(=O)O)c([2 H])c(Cl)c1Cl
IUPAC navn 3,4-dichloro-2,5,6-trideuteriobenzoic acid
Molekylvægt (g/mol) 194.03
Formel vægt 192.9777 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity