Learn More
3,4-Dichloroaniline, TRC
Product Code. 18050230
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18050230 5 g 5g
30137012 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 18050230 Supplier Toronto Research Chemicals Supplier No. TRCD4318005G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-76-1
Molecular Formula C6 H5 Cl2 N
Molecular Weight (g/mol) 162.02
Synonym Aniline, 3,4-dichloro- (7CI,8CI), 3,4-Dichlorobenzenamine, 3,4-DCA, 3,4-Dichloraniline, 3,4-Dichloroaniline, 3,4-Dichlorophenylamine, 4,5-Dichloroaniline, 4-Amino-1,2-dichlorobenzene, DCA, DCA (amine), LY 004892, NSC 247, m,p-Dichloroaniline
IUPAC Name 3,4-dichloroaniline
SMILES Nc1ccc(Cl)c(Cl)c1
Specifications
Analyte or Component Names 3,4-Dichloroaniline
Shipping Condition Room Temperature
InChI Formula InChI=1S/C6H5Cl2N/c7-5-2-1-4(9)3-6(5)8/h1-3H,9H2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 5 g
Formula Weight 160.9799
Chemical Name or Material 3,4-Dichloroaniline