Få mere at vide
3,4-Dibenzyloxyphenylacetic Acid, TRC
Artikelnummer. 30360307
Skift visning
Mængde:
1 g
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30360307 2.5 g 2.5g
30363857 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30360307 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4180552.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dibenzyloxyphenylacetic Acid
CAS 1699-61-2
Molekylær formel C22H20O4
Mængde 2.5 g
Synonym [3,4-Bis(benzyloxy)phenyl]acetic Acid, 3,4-Bis(phenylmethoxy)benzeneacetic Acid
InChI formel InChI=1S/C22H20O4/c23-22(24)14-19-11-12-20(25-15-17-7-3-1-4-8-17)21(13-19)26-16-18-9-5-2-6-10-18/h1-13H,14-16H2,(H,23,24)
SMIL OC(CC1=CC(OCC2=CC=CC=C2)=C(OCC3=CC=CC=C3)C=C1)=O
IUPAC navn 2-(3,4-bis(benzyloxy)phenyl)acetic acid
Molekylvægt (g/mol) 348.39
Formel vægt 348.14