Få mere at vide
3',4'-Dibenzyloxyaceto-d3-phenone, CDN
Artikelnummer. 30320227
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320227 1 g
30314416 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320227 Leverandør CDN Leverandørnr. CDND22771G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[3,4-Bis(phenylmethoxy)phenyl]ethanone-2,2,2-D3
CAS 344299-53-2
Molekylær formel C22 D3 H17 O3
Mængde 1 g
Synonym Ethanone-2,2,2-d3, 1-[3,4-bis(phenylmethoxy)phenyl]- (9 CI), 3',4'-Dibenzyloxyaceto-d3-phenone, 1-[3,4-Bis(phenylmethoxy)phenyl]ethanone-2,2,2-D3, 1-[3,4-Bis(benzyloxy)phenyl]ethan-1-one-2,2,2-d3, 3,4-Bis(benzyloxy)acetophenone-d3, 3',4'-Dibenzyloxyacetophenone-2,2,2-d3
InChI formel InChI=1 S/C22H20O3/c1-17(23)20-12-13-21(24-15-18-8-4-2-5-9-18)22(14-20)25-16-19-10-6-3-7-11-19/h2-14 H,15-16H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)c1ccc(OCc2ccccc2)c(OCc3ccccc3)c1
IUPAC navn 1-[3,4-bis(phenylmethoxy)phenyl]-2,2,2-trideuterioethanone
Molekylvægt (g/mol) 335.4109
Formel vægt 335.1601 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity