missing translation for 'onlineSavingsMsg'
Få mere at vide

3,4-Diaminobenzonitrile, TRC

Artikelnummer. 30358876
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358876 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30358876 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41622010G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3,4-Diaminobenzonitrile
CAS 17626-40-3
Molekylær formel C7 H7 N3
Mængde 10 g
InChI formel InChI=1S/C7H7N3/c8-4-5-1-2-6(9)7(10)3-5/h1-3H,9-10H2
SMIL Nc1ccc(cc1N)C#N
IUPAC navn 3,4-diaminobenzonitrile
Molekylvægt (g/mol) 133.15
Formel vægt 133.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.