missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diacetoxybenzamidoxime, 97%, Thermo Scientific™

Catalog Number p-7036276
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1858264-20-6
C11H12N2O5
252.23
MFCD20265286
DUQSSXZZHVNOOD-UHFFFAOYSA-N
121235395
[2-acetyloxy-4-[(Z)-N'-hydroxycarbamimidoyl]phenyl] acetate
CC(=O)OC1=C(OC(C)=O)C=C(C=C1)C(N)=NO
This item is not returnable. View return policy
1858264-20-6
C11H12N2O5
252.23
MFCD20265286
DUQSSXZZHVNOOD-UHFFFAOYSA-N
121235395
[2-acetyloxy-4-[(Z)-N'-hydroxycarbamimidoyl]phenyl] acetate
CC(=O)OC1=C(OC(C)=O)C=C(C=C1)C(N)=NO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 252.23
Percent Purity 97%
Chemical Name or Material 3,4-Diacetoxybenzamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'