Få mere at vide
3,4-Dehydrocilostazol (6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)-butoxy]-1H-quinolin-2-one), Mikromol™
Artikelnummer. 30051581
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051581 100 mg
30050277 25 mg
30459119 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051581 Leverandør Mikromol Leverandørnr. MM0796.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3,4-Dehydrocilostazol (6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)-butoxy]-1H-quinolin-2-one)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4-Dehydro Cilostazol
CAS 73963-62-9
Analyt- eller komponentnavne 3,4-Dehydro Cilostazol
Forsendelsestilstand Room Temperature
Molekylær formel C20 H25 N5 O2
Certificeringer/compliance ISO 17025
Synonym 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-2(1H)-quinolinone, 3,4-Dehydrocilostazol, OPC 13015, Cilostazol Related Compound B
InChI formel InChI=1S/C20H25N5O2/c26-20-12-9-15-14-17(10-11-18(15)21-20)27-13-5-4-8-19-22-23-24-25(19)16-6-2-1-3-7-16/h9-12,14,16H,1-8,13H2,(H,21,26)
SMIL O=C1Nc2ccc(OCCCCc3nnnn3C4CCCCC4)cc2C=C1
IUPAC navn 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-1H-quinolin-2-one
Molekylvægt (g/mol) 367.44
Mængde 100 mg
Formel vægt 367.2008 g/mol