missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chloro-2-fluoro-3-methoxyphenyl)propionic acid, 97%, Thermo Scientific™

Catalog Number p-7052457
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-13-7
C10H10ClFO3
232.635
MFCD19687213
MJIGCENLUSLAFF-UHFFFAOYSA-N
3-4-chloro-2-fluoro-3-methoxyphenyl propionic acid, 3-4-chloro-2-fluoro-3-methoxyphenyl propanoic acid
66523555
3-(4-chloro-2-fluoro-3-methoxyphenyl)propanoic acid
COC1=C(C=CC(=C1F)CCC(=O)O)Cl
This item is not returnable. View return policy
1323966-13-7
C10H10ClFO3
232.635
MFCD19687213
MJIGCENLUSLAFF-UHFFFAOYSA-N
3-4-chloro-2-fluoro-3-methoxyphenyl propionic acid, 3-4-chloro-2-fluoro-3-methoxyphenyl propanoic acid
66523555
3-(4-chloro-2-fluoro-3-methoxyphenyl)propanoic acid
COC1=C(C=CC(=C1F)CCC(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 232.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(4-Chloro-2-fluoro-3-methoxyphenyl)propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'