missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)-9-phenylcarbazole, 98%, Thermo Scientific™

Catalog Number 15491809
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1028647-93-9
C24H16BrN
398.303
MFCD14582939
JEYLGFCAZBGCMC-UHFFFAOYSA-N
3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole
51358293
3-(4-bromophenyl)-9-phenylcarbazole
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52
This item is not returnable. View return policy
1028647-93-9
C24H16BrN
398.303
MFCD14582939
JEYLGFCAZBGCMC-UHFFFAOYSA-N
3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole
51358293
3-(4-bromophenyl)-9-phenylcarbazole
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 19325695
Formula Weight 398.3
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-(4-Bromophenyl)-9-phenylcarbazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'