Få mere at vide
[3,4-Bis(benzyloxy)phenyl]-1,2-ethanediol-d5, TRC
Artikelnummer. 30336687
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30336687 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336687 Leverandør Toronto Research Chemicals Leverandørnr. TRCB41100225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale [3,4-Bis(benzyloxy)phenyl]-1,2-ethanediol-d5
CAS 1794960-44-3
Molekylær formel C22H17D5O4
Mængde 25 mg
Synonym 1-[3,4-Bis(phenylmethoxy)phenyl]-1,2-ethanediol-d5, rac 3,4-Bis(benzyloxy)phenylethylene Glycol-d5
InChI formel InChI=1S/C22H22O4/c23-14-20(24)19-11-12-21(25-15-17-7-3-1-4-8-17)22(13-19)26-16-18-9-5-2-6-10-18/h1-13,20,23-24H,14-16H2/i11D,12D,13D,14D2
SMIL [2H]c1c([2H])c(C(O)C([2H])([2H])O)c([2H])c(OCc2ccccc2)c1OCc3ccccc3
IUPAC navn 1,1-dideuterio-2-[2,3,6-trideuterio-4,5-bis(phenylmethoxy)phenyl]ethane-1,2-diol
Molekylvægt (g/mol) 355.44
Formel vægt 355.1832