missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Biphenylyl)-N-Fmoc-D-alanine, 95%, Thermo Scientific Chemicals

Catalog Number p-7034423
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
205526-38-1
C30H25NO4
463.53
MFCD00191199
VSGACONKQRJFGX-UHFFFAOYNA-N
fmoc-d-4,4'-biphenylalanine, fmoc-d-bip-oh, fmoc-d-bip 4,4'-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid, fmoc-4-biphenyl-d-ala, fmoc-l-phe 4-ph-oh, fmoc-bip-oh, 2r-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1664, fmoc-d-bip 4,4atrade mark-oh
7019142
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
OC(=O)C(CC1=CC=C(C=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
205526-38-1
C30H25NO4
463.53
MFCD00191199
VSGACONKQRJFGX-UHFFFAOYNA-N
fmoc-d-4,4'-biphenylalanine, fmoc-d-bip-oh, fmoc-d-bip 4,4'-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid, fmoc-4-biphenyl-d-ala, fmoc-l-phe 4-ph-oh, fmoc-bip-oh, 2r-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1664, fmoc-d-bip 4,4atrade mark-oh
7019142
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
OC(=O)C(CC1=CC=C(C=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 463.52
Percent Purity 95%
Chemical Name or Material 3-(4-Biphenylyl)-N-Fmoc-D-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'