Få mere at vide
3,4,5-Trimethoxybenzaldehyde-d3, TRC
Artikelnummer. 30456634
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30456634 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456634 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79563810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4,5-Trimethoxybenzaldehyde 4-methoxy-d3
CAS 1219805-17-0
Molekylær formel C10 2H3 H9 O4
Mængde 10 mg
Synonym Benzaldehyde','3,5-dimethoxy-4-(methoxy-d3)-, 3,5-Dimethoxy-4-(trideuteriomethoxy)benzaldehyde, 3,5-Dimethoxy-4-methoxy-d3-benzaldehyde, 3,4,5-Trimethoxybenzaldehyde-d3, 3,4,5-Trimethoxybenzaldehyde-D3 (4-methoxy-D3)
InChI formel InChI=1S/C10H12O4/c1-12-8-4-7(6-11)5-9(13-2)10(8)14-3/h4-6H,1-3H3/i3D3
SMIL [2H]C([2H])([2H])Oc1c(OC)cc(C=O)cc1OC
IUPAC navn 3,5-dimethoxy-4-(trideuteriomethoxy)benzaldehyde
Molekylvægt (g/mol) 199.22
Formel vægt 199.0924