Få mere at vide
3,4,5-Trimethoxy-d9-benzoic Acid, CDN
Artikelnummer. 30323615
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323615 0.5 g 0.5g
30327122 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323615 Leverandør CDN Leverandørnr. CDND76610.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4,5-Trimethoxybenzoic Acid-d9
CAS 84759-05-7
Molekylær formel C10 D9 H3 O5
Mængde 0.5 g
Synonym Benzoic acid, 3,4,5-tri(methoxy-d3)- (9 CI), 3,4,5-Tri(methoxy-d3)benzoic acid, 3,4,5-Trimethoxybenzoic acid-d9, 3,4,5-Trimethoxybenzoic Acid-D9 (trimethoxy-D9), Gallic acid tri(methyl-d3) ether, Tri(methyl-d3)gallic acid, 3,4,5-Trimethoxy-d9-benzoic Acid
InChI formel InChI=1 S/C10H12O5/c1-13-7-4-6(10(11)12)5-8(14-2)9(7)15-3/h4-5 H,1-3H3,(H,11,12)/i1D3,2D3,3D3
SMIL [2 H]C([2 H])([2 H])Oc1cc(cc(OC([2 H])([2 H])[2 H])c1OC([2 H])([2 H])[2 H])C(=O)O
IUPAC navn 3,4,5-tris(trideuteriomethoxy)benzoic acid
Molekylvægt (g/mol) 221.255
Formel vægt 221.125 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity