Få mere at vide
3,4',5-Trichlorobiphenyl-2',3',5',6'-d4, CDN
Artikelnummer. 30319460
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30319460 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319460 Leverandør CDN Leverandørnr. CDND70450.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4',5-Trichlorobiphenyl-2',3',5',6'-D4
CAS 1276197-48-8
Molekylær formel C12 D4 H3 Cl3
Mængde 0.01 g
Synonym 3,4',5-Trichlorobiphenyl-2',3',5',6'-d4, 3,4',5-Trichloro-1,1'-biphenyl-2',3',5',6'-d4, 3,4',5-Trichlorobiphenyl-2',3',5',6'-D4, PCB 39-d4, 3,4',5-TriCB-2',3',5',6'-d4
InChI formel InChI=1 S/C12H7Cl3/c13-10-3-1-8(2-4-10)9-5-11(14)7-12(15)6-9/h1-7 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Cl)c2cc(Cl)cc(Cl)c2
IUPAC navn 1-chloro-2,3,5,6-tetradeuterio-4-(3,5-dichlorophenyl)benzene
Molekylvægt (g/mol) 261.5676
Formel vægt 259.9864 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity