Få mere at vide
3,4,5-Trichlorobiphenyl-2',3',4',5',6'-d5, CDN
Artikelnummer. 30325747
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30325747 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325747 Leverandør CDN Leverandørnr. CDND30120.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,4,5-Trichlorobiphenyl-2',3',4',5',6'-D5
CAS 93952-22-8
Molekylær formel C12 D5 H2 Cl3
Mængde 0.01 g
Synonym 1,1'-Biphenyl-2,3,4,5,6-d5, 3',4',5'-trichloro- (9 CI), 3',4',5'-Trichloro-1,1'-biphenyl-2,3,4,5,6-d5, 3,4,5-Trichloro-1,1'-biphenyl-2',3',4',5',6'-d5, 3,4,5-Trichlorobiphenyl-2',3',4',5',6'-D5, PCB 38-d5, 3,4,5-TriCB-2',3',4',5',6'-d5
InChI formel InChI=1 S/C12H7Cl3/c13-10-6-9(7-11(14)12(10)15)8-4-2-1-3-5-8/h1-7 H/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2cc(Cl)c(Cl)c(Cl)c2
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(3,4,5-trichlorophenyl)benzene
Molekylvægt (g/mol) 262.5738
Formel vægt 260.9927 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity