Få mere at vide
3-[4-(2-Pyridylsulfamoyl)phenyl] Sulfasalazine, TRC
Artikelnummer. 30412405
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412405 1 mg
30427442 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30412405 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9958101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391062-34-2
Molekylær formel C29H22N6O7S2
Molekylvægt (g/mol) 630.65
Synonym 2-Hydroxy-3-[4-[(2-pyridinylamino)sulfonyl]phenyl]-5-[[4-[(2-pyridinylamino)sulfonyl]phenyl]azo]benzoic Acid, Sulfasalazine Impurity E, 3-[p-(2-Pyridylsulfamoyl)phenyl]-5-[[p-(2-Pyridylsulfamoyl)phenyl]azo]salicylic Acid
SMIL C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)C3=C/C(=N/NC4=CC=C(C=C4)S(=O)(=O)NC5=CC=CC=N5)/C=C(C3=O)C(=O)O
Tekniske data
Kemisk navn eller materiale 3-[4-(2-Pyridylsulfamoyl)phenyl] Sulfasalazine
Mængde 1 mg
InChI formel InChI=1S/C29H22N6O7S2/c36-28-24(19-7-11-22(12-8-19)43(39,40)34-26-5-1-3-15-30-26)17-21(18-25(28)29(37)38)33-32-20-9-13-23(14-10-20)44(41,42)35-27-6-2-4-16-31-27/h1-18,36H,(H,30,34)(H,31,35)(H,37,38)
Formel vægt 630.0991