missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[3-(Trifluoromethyl)phenyl]propionic acid, 97%, Alfa Aesar™
Brand: Alfa Aesar H32088.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
585-50-2 | |
218.175 | |
YLTJJMIWCCJIHI-UHFFFAOYSA-N | |
2777683 | |
C1=CC(=CC(=C1)C(F)(F)F)CCC(=O)O |
C10H9F3O2 | |
MFCD00090487 | |
3-3-trifluoromethyl phenyl propanoic acid, 3-3-trifluoromethylphenyl propionic acid, benzenepropanoic acid, 3-trifluoromethyl, 3-trifluoromethyl hydrocinnamic acid, unii-4u44xhh79h, 3-3'-trifluoromethylphenyl-propionic acid, 3-3-trifluoromethyl phenyl propionic acid, cinacalcet m5, m-trifluoromethyl hydrocinnamic acid | |
3-[3-(trifluoromethyl)phenyl]propanoic acid |
Specifications
3-[3-(Trifluoromethyl)phenyl]propionic acid | |
1g | |
C10H9F3O2 | |
3-3-trifluoromethyl phenyl propanoic acid, 3-3-trifluoromethylphenyl propionic acid, benzenepropanoic acid, 3-trifluoromethyl, 3-trifluoromethyl hydrocinnamic acid, unii-4u44xhh79h, 3-3'-trifluoromethylphenyl-propionic acid, 3-3-trifluoromethyl phenyl propionic acid, cinacalcet m5, m-trifluoromethyl hydrocinnamic acid | |
C1=CC(=CC(=C1)C(F)(F)F)CCC(=O)O | |
218.175 | |
218.17 |
34°C to 37°C | |
585-50-2 | |
MFCD00090487 | |
YLTJJMIWCCJIHI-UHFFFAOYSA-N | |
3-[3-(trifluoromethyl)phenyl]propanoic acid | |
2777683 | |
97% |