Learn More
3-(3-Thienyl)acrylic Acid, TRC
Product Code. 30454718
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30454718 500 mg 500mg
1 options
Product Code. 30454718 Supplier Toronto Research Chemicals Supplier No. TRCT430030500MG
 Restricted Item
Forventet forsendelse: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(3-Thienyl)acrylic Acid
CAS 102696-71-9
Molecular Formula C7H6O2S
Quantity 500 mg
Synonym (2E)-3-(3-Thienyl)-2-propenoic Acid, (E)-3-(3-Thienyl)-2-propenoic Acid, (E)-3-(Thiophen-3-yl)acrylic Acid, trans-3-(3-Thienyl)acrylic Acid, trans-3-(Thiophen-3-yl)acrylic Acid
InChI Formula InChI=1S/C7H6O2S/c8-7(9)2-1-6-3-4-10-5-6/h1-5H,(H,8,9)/b2-1+
SMILES OC(=O)\C=C\c1ccsc1
IUPAC Name (E)-3-thiophen-3-ylprop-2-enoic acid
Molecular Weight (g/mol) 154.19
Formula Weight 154.0089