Få mere at vide
3,3'-Sulfonylbis(N-(4-cyano-3-(trifluoromethyl)phenyl)-2-hydroxy-2-methylpropanamide)(Mixture of Diastereomers), TRC
Artikelnummer. 30135858
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135858 100 mg
30009206 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135858 Leverandør Toronto Research Chemicals Leverandørnr. TRCS699335100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2711603-77-7
Molekylær formel C24 H20 F6 N4 O6 S
Molekylvægt (g/mol) 606.494
Synonym 3,3'-Sulfonylbis[N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide]
IUPAC navn 3-[3-[4-cyano-3-(trifluoromethyl)anilino]-2-hydroxy-2-methyl-3-oxopropyl]sulfonyl-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide
SMIL CC(O)(CS(=O)(=O)CC(C)(O)C(=O)Nc1ccc(C#N)c(c1)C(F)(F)F)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 3,3'-Sulfonylbis[N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide]
Mængde 100 mg
InChI formel InChI=1S/C24H20F6N4O6S/c1-21(37,19(35)33-15-5-3-13(9-31)17(7-15)23(25,26)27)11-41(39,40)12-22(2,38)20(36)34-16-6-4-14(10-32)18(8-16)24(28,29)30/h3-8,37-38H,11-12H2,1-2H3,(H,33,35)(H,34,36)
Formel vægt 606.101