missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055378
355.23 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
64090-98-8
C8H10N2O2
166.18
MFCD00079676
DFZVZEMNPGABKO-ZETCQYMHSA-N
3-3-pyridyl-l-alanine, l-3-pyridylalanine, s-2-amino-3-pyridin-3-yl propanoic acid, h-3-pal-oh, 3-3-pyridyl alanine, 2s-2-amino-3-pyridin-3-yl propanoic acid, l-3-3-pyridyl-alanine, 3'-pyridyl-l-ala, h-beta-3-pyridyl-ala-oh, 2s-2-amino-3-pyridin-3-ylpropanoic acid
152953
(2S)-2-amino-3-pyridin-3-ylpropanoic acid
C1=CC(=CN=C1)CC(C(=O)O)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
64090-98-8
C8H10N2O2
166.18
MFCD00079676
DFZVZEMNPGABKO-ZETCQYMHSA-N
3-3-pyridyl-l-alanine, l-3-pyridylalanine, s-2-amino-3-pyridin-3-yl propanoic acid, h-3-pal-oh, 3-3-pyridyl alanine, 2s-2-amino-3-pyridin-3-yl propanoic acid, l-3-3-pyridyl-alanine, 3'-pyridyl-l-ala, h-beta-3-pyridyl-ala-oh, 2s-2-amino-3-pyridin-3-ylpropanoic acid
152953
(2S)-2-amino-3-pyridin-3-ylpropanoic acid
C1=CC(=CN=C1)CC(C(=O)O)N

3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 248°C to 252°C (decomposition)
Color White
Beilstein 4351583
Solubility Information Soluble in clear solution (0.3 gram in 2 ml DMF).
Formula Weight 166.18
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material 3-(3-Pyridyl)-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'