Learn More
Alfa Aesar™ 3-(3-Pyridyl)benzeneboronic acid, 98%
Brand:  Alfa Aesar™ H64739.MD
Additional Details : CAS Number : 351422-72-5 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15472049
  • 181.23 DKK / 250mg
Estimated Shipment
09-09-2020
Add to basket
Description
Description
Applications
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Chemical Identifiers
351422-72-5
199.016
SSVQXHWHJJERAP-UHFFFAOYSA-N
58526507
B(C1=CC(=CC=C1)C2=CN=CC=C2)(O)O
C11H10BNO2
MFCD16658918
3-pyridin-3-yl phenyl boronic acid, 3-3-pyridyl benzeneboronic acid, 3-pyridin-3-yl phenylboronic acid, 3-3-pyridinyl phenylboronic acid, 3-3-pyridinyl phenyl boronic acid, boronic acid, 3-3-pyridinyl phenyl, 3-3-pyridyl phenylboronic acid, 3-pyridin-3-yl phenyl boronic acid pinacol ester
(3-pyridin-3-ylphenyl)boronic acid
Specifications
3-(3-Pyridyl)benzeneboronic acid
250mg
C11H10BNO2
20520996
SSVQXHWHJJERAP-UHFFFAOYSA-N
(3-pyridin-3-ylphenyl)boronic acid
58526507
Powder
White
351422-72-5
MFCD16658918
3-pyridin-3-yl phenyl boronic acid, 3-3-pyridyl benzeneboronic acid, 3-pyridin-3-yl phenylboronic acid, 3-3-pyridinyl phenylboronic acid, 3-3-pyridinyl phenyl boronic acid, boronic acid, 3-3-pyridinyl phenyl, 3-3-pyridyl phenylboronic acid, 3-pyridin-3-yl phenyl boronic acid pinacol ester
B(C1=CC(=CC=C1)C2=CN=CC=C2)(O)O
199.016
199.02
98%
Videos
SDS Documents