missing translation for 'onlineSavingsMsg'
Learn More

3-[3-Fluoro-5-(trifluoromethoxy)phenyl]propionic acid, 97%, Thermo Scientific™

Catalog Number 15488835
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1261828-37-8
C10H8F4O3
252.165
MFCD18783232
OXUSROHJYYUZSB-UHFFFAOYSA-N
3-3-fluoro-5-trifluoromethoxy phenyl propionic acid, 3-3-fluoro-5-trifluoromethoxy phenyl propanoic acid
66523563
3-[3-fluoro-5-(trifluoromethoxy)phenyl]propanoic acid
C1=C(C=C(C=C1OC(F)(F)F)F)CCC(=O)O
This item is not returnable. View return policy
1261828-37-8
C10H8F4O3
252.165
MFCD18783232
OXUSROHJYYUZSB-UHFFFAOYSA-N
3-3-fluoro-5-trifluoromethoxy phenyl propionic acid, 3-3-fluoro-5-trifluoromethoxy phenyl propanoic acid
66523563
3-[3-fluoro-5-(trifluoromethoxy)phenyl]propanoic acid
C1=C(C=C(C=C1OC(F)(F)F)F)CCC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 252.16
Percent Purity 97%
Chemical Name or Material 3-[3-Fluoro-5-(trifluoromethoxy)phenyl]propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'