missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-(3-Fluoro-5-methylphenyl)propionic acid, 97%
Brand: Alfa Aesar™ H32551.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1017779-56-4 | |
182.194 | |
SEUXGGXOPIWHGW-UHFFFAOYSA-N | |
46737683 | |
CC1=CC(=CC(=C1)F)CCC(=O)O |
C10H11FO2 | |
MFCD09832369 | |
3-3-fluoro-5-methylphenyl propanoic acid, 3-3-fluoro-5-methylphenyl propionic acid, 3-3-fluoro-5-methylphenyl propionic acid, jrd | |
3-(3-fluoro-5-methylphenyl)propanoic acid |
Specifications
3-(3-Fluoro-5-methylphenyl)propionic acid | |
1g | |
C10H11FO2 | |
3-3-fluoro-5-methylphenyl propanoic acid, 3-3-fluoro-5-methylphenyl propionic acid, 3-3-fluoro-5-methylphenyl propionic acid, jrd | |
CC1=CC(=CC(=C1)F)CCC(=O)O | |
182.194 | |
182.19 |
64°C to 66°C | |
1017779-56-4 | |
MFCD09832369 | |
SEUXGGXOPIWHGW-UHFFFAOYSA-N | |
3-(3-fluoro-5-methylphenyl)propanoic acid | |
46737683 | |
97% |