Få mere at vide
3-(3-Ethoxy-4-methoxyphenyl)propionic-2,2,3,3-d4 Acid, CDN
Artikelnummer. 30322678
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322678 0.1 g
30314678 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322678 Leverandør CDN Leverandørnr. CDND74200.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(3-Ethoxy-4-methoxyphenyl)propanoic Acid-2,2,3,3-D4
CAS 1398065-51-4
Molekylær formel C12 D4 H12 O4
Mængde 0.1 g
Synonym Benzenepropanoic-α,α,β,β-d4 acid, 3-ethoxy-4-methoxy- (ACI), 3-Ethoxy-4-methoxybenzenepropanoic-α,α,β,β-d4 acid (ACI), 3-(3-Ethoxy-4-methoxyphenyl)propionic-2,2,3,3-d4 Acid, 3-(3-Ethoxy-4-methoxyphenyl)propanoic Acid-2,2,3,3-D4, 3-Ethoxy-4-methoxyhydrocinnamic-d4 acid
InChI formel InChI=1 S/C12H16O4/c1-3-16-11-8-9(5-7-12(13)14)4-6-10(11)15-2/h4,6,8 H,3,5,7H2,1-2H3,(H,13,14)/i5D2,7D2
SMIL [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])c1ccc(OC)c(OCC)c1
IUPAC navn 2,2,3,3-tetradeuterio-3-(3-ethoxy-4-methoxyphenyl)propanoic acid
Molekylvægt (g/mol) 228.2777
Formel vægt 228.13 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity