missing translation for 'onlineSavingsMsg'
Få mere at vide

3,3'-Dithio(succinimidyl propionate), TRC

Artikelnummer. 30380845
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30380845 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30380845 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49427525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3,3'-Dithio(succinimidyl propionate)
CAS 57757-57-0
Molekylær formel C14H16N2O8S2
Mængde 25 mg
Synonym 1,1'-[Dithiobis[(1-oxo-3,1-propanediyl)oxy]]bis-2,5-pyrrolidinedione, Bis[2-[(succinimidyloxy)carbonyl]ethyl] disulfide, DTSP, Lomant's Reagent, NSC 328386
InChI formel InChI=1S/C14H16N2O8S2/c17-9-1-2-10(18)15(9)23-13(21)5-7-25-26-8-6-14(22)24-16-11(19)3-4-12(16)20/h1-8H2
SMIL O=C(CCSSCCC(=O)ON1C(=O)CCC1=O)ON2C(=O)CCC2=O
IUPAC navn (2,5-dioxopyrrolidin-1-yl) 3-[[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]disulfanyl]propanoate
Molekylvægt (g/mol) 404.42
Formel vægt 404.0348
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.