missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethylglutarimide, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Beschreibung
3,3'-dimethylglutarimide were used as the barbital subsitutes. 3,3-dimethylglutarimide is involved formation of the heterodimer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′-dimethylglutarimide were used as the barbital subsitutes. 3,3-dimethylglutarimide is involved formation of the heterodimer.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
3,3′-dimethylglutarimide were used as the barbital subsitutes. 3,3-dimethylglutarimide is involved formation of the heterodimer.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Spezifikation
Melting Point | 144°C to 146°C |
Quantity | 5 g |
Beilstein | 111981 |
Solubility Information | Soluble in water. |
Formula Weight | 141.17 |
Percent Purity | 99% |
Chemical Name or Material | 3,3-Dimethylglutarimide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'